Product Details
- SNP ID
-
rs1049867
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:14882019 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAAAGGGGTGCAGCCAGACAAGA[A/G]AATATACAGGAAAGAAGCATTGTAT
- Phenotype
-
MIM: 154870
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MGP
PubMed Links
Gene Details
- Gene
- MGP
- Gene Name
- matrix Gla protein
View Full Product Details