Product Details

SNP ID
rs1599965
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:95408070 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACATTCTCAGCATCGCCCCAGACA[C/T]CCTTGTTCACAATCTCAGGGGTGGA
Phenotype
MIM: 610861
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00341 PubMed Links

Gene Details

Gene
LINC00341
Gene Name
long intergenic non-protein coding RNA 341
There are no transcripts associated with this gene.

Gene
SYNE3
Gene Name
spectrin repeat containing nuclear envelope family member 3
There are no transcripts associated with this gene.

View Full Product Details