Product Details

SNP ID
rs1061220
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:143426133 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TATCTCACCACTTTCAGAACTATTT[C/T]GTACACAAAACAAGTCTTTCAGCTG
Phenotype
MIM: 615388
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADAT2 PubMed Links

Gene Details

Gene
ADAT2
Gene Name
adenosine deaminase, tRNA specific 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286259.1 2830 UTR 3 NP_001273188.1
NM_182503.2 2830 UTR 3 NP_872309.2
XM_011535439.2 2830 UTR 3 XP_011533741.1
XM_011535440.1 2830 UTR 3 XP_011533742.1
XM_011535443.1 2830 Intron XP_011533745.1
XM_011535444.2 2830 UTR 3 XP_011533746.1
XM_017010260.1 2830 UTR 3 XP_016865749.1
XM_017010261.1 2830 UTR 3 XP_016865750.1
XM_017010262.1 2830 UTR 3 XP_016865751.1
XM_017010263.1 2830 UTR 3 XP_016865752.1

View Full Product Details