Product Details

SNP ID
rs941609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:49990011 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTATTGTTTGACTTTGCCTTGTTT[C/G]TGCCTATGAATTACTAACAGAGGCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LINC01588 PubMed Links

Gene Details

Gene
LINC01588
Gene Name
long intergenic non-protein coding RNA 1588
There are no transcripts associated with this gene.

View Full Product Details