Product Details

SNP ID
rs1187877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:55028443 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTTTTTGGAGATCATTTGTTT[C/T]TTGGATAGACTGGCTTATAAATTTT
Phenotype
MIM: 616337 MIM: 608126
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SOCS4 PubMed Links

Gene Details

Gene
SOCS4
Gene Name
suppressor of cytokine signaling 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080867.2 Intron NP_543143.1
NM_199421.1 Intron NP_955453.1
XM_011536425.1 Intron XP_011534727.1
XM_011536426.1 Intron XP_011534728.1
Gene
WDHD1
Gene Name
WD repeat and HMG-box DNA binding protein 1
There are no transcripts associated with this gene.

View Full Product Details