Product Details

SNP ID
rs1254331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:60449519 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTTATCATGCCCTTTATTACTCTA[A/G]CCTTATAGGATGTTTGAATATTTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C14orf39 PubMed Links

Gene Details

Gene
C14orf39
Gene Name
chromosome 14 open reading frame 39
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174978.2 Intron NP_777638.2
XM_011536690.2 Intron XP_011534992.1
XM_011536703.2 Intron XP_011535005.1
XM_011536704.1 Intron XP_011535006.1
XM_017021247.1 Intron XP_016876736.1
XM_017021248.1 Intron XP_016876737.1
XM_017021249.1 Intron XP_016876738.1
XM_017021250.1 Intron XP_016876739.1
XM_017021251.1 Intron XP_016876740.1

View Full Product Details