Product Details

SNP ID
rs1801077
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2255121 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTCAGGGTGCTGTGTGGTCCCG[C/G]AAAGAGGCAGCTGAGCTTGGGCCTC
Phenotype
MIM: 600312 MIM: 614905
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MIR6836 PubMed Links

Gene Details

Gene
MIR6836
Gene Name
microRNA 6836
There are no transcripts associated with this gene.

Gene
NUDT1
Gene Name
nudix hydrolase 1
There are no transcripts associated with this gene.

Gene
SNX8
Gene Name
sorting nexin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013321.3 1499 Missense Mutation CCG,GCG P445A NP_037453.1
XM_011515329.2 1499 Missense Mutation CCG,GCG P437A XP_011513631.1
XM_011515330.1 1499 Missense Mutation CCG,GCG P431A XP_011513632.1
XM_017012083.1 1499 Missense Mutation CCG,GCG P392A XP_016867572.1
XM_017012084.1 1499 Missense Mutation CCG,GCG P392A XP_016867573.1

View Full Product Details