Product Details

SNP ID
rs1359701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:12777127 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTTCTATCACTCCGACAGGAATG[A/C]TGGAAATTATTTCTATCATTCTATC
Phenotype
MIM: 616130
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LURAP1L PubMed Links

Gene Details

Gene
LURAP1L
Gene Name
leucine rich adaptor protein 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203403.1 Intron NP_981948.1
XM_005251443.2 Intron XP_005251500.1
Gene
LURAP1L-AS1
Gene Name
LURAP1L antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details