Product Details

SNP ID
rs1022654
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:59934882 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGATCAAATCACAGTAAATAAGCT[A/C]TCAGAGGTGTTTTAAATATTCTGCT
Phenotype
MIM: 603326 MIM: 604105
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM217B PubMed Links

Gene Details

Gene
FAM217B
Gene Name
family with sequence similarity 217 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190826.1 Intron NP_001177755.1
NM_001190827.1 Intron NP_001177756.1
NM_022106.2 Intron NP_071389.1
XM_011528985.2 Intron XP_011527287.1
XM_011528986.2 Intron XP_011527288.1
Gene
PPP1R3D
Gene Name
protein phosphatase 1 regulatory subunit 3D
There are no transcripts associated with this gene.

Gene
SYCP2
Gene Name
synaptonemal complex protein 2
There are no transcripts associated with this gene.

View Full Product Details