Product Details

SNP ID
hCV7613631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:41619390 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAAATGTTTGCATCAGAGTATAG[A/C]AATTCCTGTAAAAACAGAGAAGAGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CEACAM4 PubMed Links

Gene Details

Gene
CEACAM4
Gene Name
carcinoembryonic antigen related cell adhesion molecule 4
There are no transcripts associated with this gene.

View Full Product Details