Product Details
- SNP ID
-
rs12108
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:44983904 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CAGGTCCCCTCTAGCTCCAGCATCT[A/T]GGGATTGCTGCTGTAGAAACACTGA
- Phenotype
-
MIM: 600642
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MYO1G
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80217205] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MYO1G
- Gene Name
- myosin IG
There are no transcripts associated with this gene.
- Gene
- SNHG15
- Gene Name
- small nucleolar RNA host gene 15
There are no transcripts associated with this gene.
- Gene
- SNORA9
- Gene Name
- small nucleolar RNA, H/ACA box 9
There are no transcripts associated with this gene.
View Full Product Details