Product Details

SNP ID
rs1438832
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223947235 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGATGAGATTAATAAGACATGGTT[C/T]TTGTGAGGCTGTACAGTGTAATGAC
Phenotype
MIM: 611849
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
MRPL44 PubMed Links

Gene Details

Gene
MRPL44
Gene Name
mitochondrial ribosomal protein L44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022915.3 Intron NP_075066.1
XM_011511668.2 Intron XP_011509970.1
XM_017004730.1 Intron XP_016860219.1
XM_017004731.1 Intron XP_016860220.1
Gene
WDFY1
Gene Name
WD repeat and FYVE domain containing 1
There are no transcripts associated with this gene.

View Full Product Details