Product Details

SNP ID
rs849011
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:11715623 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTCACGTTGTTTTCATTGGTGTGT[C/G]TACTTCTTTCTCTTCTGTTTTTCCC
Phenotype
MIM: 614348
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADTRP PubMed Links

Gene Details

Gene
ADTRP
Gene Name
androgen dependent TFPI regulating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143948.1 Intron NP_001137420.1
NM_032744.3 Intron NP_116133.1
XM_005249454.3 Intron XP_005249511.1
XM_011514956.1 Intron XP_011513258.1
XM_011514958.2 Intron XP_011513260.1
XM_011514959.1 Intron XP_011513261.1
XM_017011377.1 Intron XP_016866866.1
Gene
LOC340184
Gene Name
uncharacterized LOC340184
There are no transcripts associated with this gene.

View Full Product Details