Product Details

SNP ID
rs1389722
Assay Type
Functionally tested
NCBI dbSNP Submissions
74
Location
Chr.1:110875595 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCAGCCTAGGGCACTTCTTGTACC[A/G]AGCAAGCATCCACTGTGTGTAGATC
Phenotype
MIM: 151525 MIM: 615354
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD53 PubMed Links

Gene Details

Gene
CD53
Gene Name
CD53 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000560.3 Intron NP_000551.1
NM_001040033.1 Intron NP_001035122.1
NM_001320638.1 Intron NP_001307567.1
Gene
LRIF1
Gene Name
ligand dependent nuclear receptor interacting factor 1
There are no transcripts associated with this gene.

View Full Product Details