Product Details

SNP ID
rs3893196
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67292461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGAACTAAGACGTATCAAATGG[A/G]GCTGAGGGCTGGAAATCTTTATAAA
Phenotype
MIM: 609526
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCTD19 PubMed Links

Gene Details

Gene
KCTD19
Gene Name
potassium channel tetramerization domain containing 19
There are no transcripts associated with this gene.

Gene
PLEKHG4
Gene Name
pleckstrin homology and RhoGEF domain containing G4
There are no transcripts associated with this gene.

View Full Product Details