Product Details

SNP ID
rs7251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49659652 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACACCATGAGGAGCGAGGGCTCAG[C/G]TCTCCCCAGGGCCCTGGAAATCCAT
Phenotype
MIM: 610837 MIM: 603734
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BCL2L12 PubMed Links

Gene Details

Gene
BCL2L12
Gene Name
BCL2 like 12
There are no transcripts associated with this gene.

Gene
IRF3
Gene Name
interferon regulatory factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001197122.1 1591 Missense Mutation GAC,GAG D432E NP_001184051.1
NM_001197123.1 1591 Missense Mutation ACC,AGC T392S NP_001184052.1
NM_001197124.1 1591 Missense Mutation ACC,AGC T300S NP_001184053.1
NM_001197125.1 1591 Missense Mutation ACC,AGC T281S NP_001184054.1
NM_001197126.1 1591 Missense Mutation ACC,AGC T281S NP_001184055.1
NM_001197127.1 1591 Missense Mutation ACC,AGC T154S NP_001184056.1
NM_001197128.1 1591 Missense Mutation ACC,AGC T154S NP_001184057.1
NM_001571.5 1591 Missense Mutation ACC,AGC T427S NP_001562.1
XM_006723197.1 1591 Missense Mutation GAC,GAG D432E XP_006723260.1
XM_006723198.1 1591 Missense Mutation GAC,GAG D432E XP_006723261.1
XM_006723200.1 1591 Missense Mutation GAC,GAG D397E XP_006723263.1
XM_006723201.1 1591 Missense Mutation GAC,GAG D286E XP_006723264.1
XM_006723202.2 1591 Missense Mutation GAC,GAG D286E XP_006723265.1
XM_017026766.1 1591 Missense Mutation ACC,AGC T427S XP_016882255.1
XM_017026767.1 1591 Missense Mutation ACC,AGC T427S XP_016882256.1
Gene
SCAF1
Gene Name
SR-related CTD associated factor 1
There are no transcripts associated with this gene.

View Full Product Details