Product Details

SNP ID
rs12462756
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:49595166 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CGGGTGCTGCCCCGGGCCCACCGCC[A/G]CCTGAGCGGGCCCTGCCACGCCAGG
Phenotype
MIM: 616633 MIM: 604429
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRR12 PubMed Links

Gene Details

Gene
PRR12
Gene Name
proline rich 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017027031.1 1309 Silent Mutation CCA,CCG P277P XP_016882520.1
Gene
PRRG2
Gene Name
proline rich and Gla domain 2
There are no transcripts associated with this gene.

View Full Product Details