Product Details

SNP ID
rs3933376
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:127970926 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATGCTACAGTGATAAACTGCAAA[A/T]GTTTTTCTCCATATATTTCAGATGA
Phenotype
MIM: 602545
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PTPRK PubMed Links

Gene Details

Gene
PTPRK
Gene Name
protein tyrosine phosphatase, receptor type K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135648.2 Intron NP_001129120.1
NM_001291981.1 Intron NP_001278910.1
NM_001291982.1 Intron NP_001278911.1
NM_001291983.1 Intron NP_001278912.1
NM_001291984.1 Intron NP_001278913.1
NM_002844.3 Intron NP_002835.2
XM_006715537.2 Intron XP_006715600.1
XM_011536014.2 Intron XP_011534316.1
XM_011536015.2 Intron XP_011534317.1
XM_011536016.2 Intron XP_011534318.1
XM_011536017.2 Intron XP_011534319.1
XM_011536018.2 Intron XP_011534320.1
XM_011536020.2 Intron XP_011534322.1
XM_011536021.2 Intron XP_011534323.1
XM_017011145.1 Intron XP_016866634.1
XM_017011146.1 Intron XP_016866635.1
XM_017011147.1 Intron XP_016866636.1
XM_017011148.1 Intron XP_016866637.1
XM_017011149.1 Intron XP_016866638.1
XM_017011150.1 Intron XP_016866639.1

View Full Product Details