Product Details
- SNP ID
-
rs12662471
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:41553715 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGAGGGAGGAAGAGCTGGGAACAG[A/T]CTGCACCCTAGTTGGGGACAGAGGA
- Phenotype
-
MIM: 608924
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
FOXP4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78182691] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FOXP4
- Gene Name
- forkhead box P4
- Gene
- FOXP4-AS1
- Gene Name
- FOXP4 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details