Product Details
- SNP ID
-
rs10901270
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:130670191 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCACTTCTCTGCTCCCCTTGGGGT[G/T]GGGCTAGGGCTTGGTGGAGATTTTT
- Phenotype
-
MIM: 616458
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PRDM12
PubMed Links
Gene Details
- Gene
- PRDM12
- Gene Name
- PR domain 12
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021619.2 |
|
Intron |
|
|
NP_067632.2 |
View Full Product Details