Product Details

SNP ID
rs35012773
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.18:3262786 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
AGCCTCCAAGGAAGCCATTAGCATG[C/T]CGGTAACCGAGTGTGTGGTTGATAC
Phenotype
MIM: 609211
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC104968399 PubMed Links

Gene Details

Gene
LOC104968399
Gene Name
uncharacterized LOC104968399
There are no transcripts associated with this gene.

Gene
MYL12A
Gene Name
myosin light chain 12A
There are no transcripts associated with this gene.

Gene
MYL12B
Gene Name
myosin light chain 12B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144944.1 Intron NP_001138416.1
NM_001144945.1 Intron NP_001138417.1
NM_033546.3 Intron NP_291024.1

View Full Product Details