Product Details

SNP ID
rs12787728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57301582 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTCCCACTGCCCCTGAGGTTCCAG[A/G]ATAAGAACACATCTTGTTCTTCTTT
Phenotype
MIM: 607104
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TNKS1BP1 PubMed Links

Gene Details

Gene
TNKS1BP1
Gene Name
tankyrase 1 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033396.2 Intron NP_203754.2
XM_006718725.3 Intron XP_006718788.1
XM_011545325.2 Intron XP_011543627.1

View Full Product Details