Product Details

SNP ID
rs1971785
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48209357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTGCAGGAAAATAGAATATAAG[C/G]CAAGAAATTTAAAATATAGAGAACA
Phenotype
MIM: 609051
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CARD8 PubMed Links

Gene Details

Gene
CARD8
Gene Name
caspase recruitment domain family member 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184900.1 4432 UTR 3 NP_001171829.1
NM_001184901.1 4432 UTR 3 NP_001171830.1
NM_001184902.1 4432 UTR 3 NP_001171831.1
NM_001184903.1 4432 UTR 3 NP_001171832.1
NM_001184904.1 4432 Intron NP_001171833.1
NM_014959.3 4432 UTR 3 NP_055774.2
XM_006723091.3 4432 UTR 3 XP_006723154.1
XM_006723092.3 4432 UTR 3 XP_006723155.1
XM_006723093.3 4432 UTR 3 XP_006723156.1
XM_006723094.3 4432 UTR 3 XP_006723157.1
XM_006723095.3 4432 UTR 3 XP_006723158.1
XM_006723096.3 4432 UTR 3 XP_006723159.1
XM_006723097.3 4432 UTR 3 XP_006723160.1
XM_006723098.3 4432 UTR 3 XP_006723161.1
XM_006723099.3 4432 UTR 3 XP_006723162.1
XM_006723101.2 4432 Intron XP_006723164.1
XM_006723102.3 4432 UTR 3 XP_006723165.1
XM_006723104.3 4432 UTR 3 XP_006723167.1
XM_006723106.3 4432 UTR 3 XP_006723169.1
XM_006723107.3 4432 UTR 3 XP_006723170.1
XM_006723109.3 4432 UTR 3 XP_006723172.1
XM_006723110.3 4432 Intron XP_006723173.1
XM_011526641.2 4432 UTR 3 XP_011524943.1
XM_011526642.2 4432 UTR 3 XP_011524944.1
XM_011526643.2 4432 UTR 3 XP_011524945.1
XM_011526644.2 4432 UTR 3 XP_011524946.1
XM_011526646.2 4432 UTR 3 XP_011524948.1
XM_011526647.2 4432 UTR 3 XP_011524949.1
XM_011526648.2 4432 UTR 3 XP_011524950.1
XM_011526650.2 4432 Intron XP_011524952.1
XM_011526653.2 4432 UTR 3 XP_011524955.1
XM_017026482.1 4432 UTR 3 XP_016881971.1
XM_017026483.1 4432 UTR 3 XP_016881972.1
XM_017026484.1 4432 UTR 3 XP_016881973.1
XM_017026485.1 4432 Intron XP_016881974.1
XM_017026486.1 4432 UTR 3 XP_016881975.1
XM_017026487.1 4432 UTR 3 XP_016881976.1
XM_017026488.1 4432 UTR 3 XP_016881977.1
XM_017026489.1 4432 Intron XP_016881978.1
XM_017026490.1 4432 Intron XP_016881979.1
XM_017026491.1 4432 Intron XP_016881980.1
XM_017026492.1 4432 UTR 3 XP_016881981.1
XM_017026493.1 4432 UTR 3 XP_016881982.1
XM_017026494.1 4432 UTR 3 XP_016881983.1
XM_017026495.1 4432 UTR 3 XP_016881984.1
XM_017026496.1 4432 UTR 3 XP_016881985.1
XM_017026497.1 4432 UTR 3 XP_016881986.1
XM_017026498.1 4432 UTR 3 XP_016881987.1
XM_017026499.1 4432 UTR 3 XP_016881988.1

View Full Product Details