Product Details

SNP ID
rs15781
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:113721674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGGAGGGAGAAAAGCTTTAAAA[A/G]AAAAGTGTTGGGGTGGGGGAGGAAT
Phenotype
MIM: 610834
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NAA50 PubMed Links

Gene Details

Gene
NAA50
Gene Name
N(alpha)-acetyltransferase 50, NatE catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308445.1 943 UTR 3 NP_001295374.1
NM_025146.3 943 UTR 3 NP_079422.1

View Full Product Details