Product Details

SNP ID
rs1128163
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121631726 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCCCCAAGCCCTTAATGAAGCAGG[A/G]GAGGGAAGTCTGTCCAGAACCTCAT
Phenotype
MIM: 609107 MIM: 601306
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO40 PubMed Links

Gene Details

Gene
FBXO40
Gene Name
F-box protein 40
There are no transcripts associated with this gene.

Gene
HCLS1
Gene Name
hematopoietic cell-specific Lyn substrate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001292041.1 1599 UTR 3 NP_001278970.1
NM_005335.5 1599 UTR 3 NP_005326.2

View Full Product Details