Product Details

SNP ID
rs793975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:121581386 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGACAAAATATAATCCAATGAGAT[A/G]GTTTACATTCTTTGTTCATAGCAAG
Phenotype
MIM: 611164
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARGFX PubMed Links

Gene Details

Gene
ARGFX
Gene Name
arginine-fifty homeobox
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012659.1 Intron NP_001012677.1

View Full Product Details