Product Details

SNP ID
rs1459189
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:64775056 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTCTTTGCTATCTTACGCTTCTT[C/T]CTTTCATTCTCTAAATTTGGGTATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CWC27 PubMed Links

Gene Details

Gene
CWC27
Gene Name
CWC27 spliceosome associated protein homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297644.1 Intron NP_001284573.1
NM_001297645.1 Intron NP_001284574.1
NM_001318000.1 Intron NP_001304929.1
NM_005869.3 Intron NP_005860.2
XM_011543095.2 Intron XP_011541397.2
XM_011543096.1 Intron XP_011541398.1
XM_017008943.1 Intron XP_016864432.1
Gene
SREK1IP1
Gene Name
SREK1 interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details