Product Details

SNP ID
rs1053867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99411108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGGAAAATGCCTAAAGTCAAA[A/C]GAAGCCGGAAAGCACCCCCAGATGG
Phenotype
MIM: 603477 MIM: 607075 MIM: 614774
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ATP5J2-PTCD1 PubMed Links

Gene Details

Gene
ATP5J2-PTCD1
Gene Name
ATP5J2-PTCD1 readthrough
There are no transcripts associated with this gene.

Gene
BUD31
Gene Name
BUD31 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003910.3 471 Silent Mutation AGA,CGA R6R NP_003901.2
XM_005250670.4 471 Silent Mutation AGA,CGA R6R XP_005250727.1
XM_005250671.4 471 Silent Mutation AGA,CGA R6R XP_005250728.1
XM_005250674.3 471 Silent Mutation AGA,CGA R6R XP_005250731.1
XM_017012760.1 471 Silent Mutation AGA,CGA R6R XP_016868249.1
XM_017012761.1 471 Silent Mutation AGA,CGA R6R XP_016868250.1
XM_017012762.1 471 Silent Mutation AGA,CGA R6R XP_016868251.1
Gene
PDAP1
Gene Name
PDGFA associated protein 1
There are no transcripts associated with this gene.

Gene
PTCD1
Gene Name
pentatricopeptide repeat domain 1
There are no transcripts associated with this gene.

View Full Product Details