Product Details

SNP ID
rs7692
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:43336269 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTGGAACAGGGAATGAGTTCAATCT[A/C]AAGACAAGTTTCAGAGATCTTCTAC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF318 PubMed Links

Gene Details

Gene
ZNF318
Gene Name
zinc finger protein 318
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014345.2 7807 UTR 3 NP_055160.2
XM_011514451.2 7807 Intron XP_011512753.1
XM_011514452.2 7807 Intron XP_011512754.1

View Full Product Details