Product Details

SNP ID
rs1129553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:103951362 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTGGCAGTTTTCAAGAAATATC[A/T]ATCAAACCGTAATTAAATTTCAACG
Phenotype
MIM: 191175
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C12orf73 PubMed Links

Gene Details

Gene
C12orf73
Gene Name
chromosome 12 open reading frame 73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135570.1 597 UTR 3 NP_001129042.1
XM_011538718.2 597 UTR 3 XP_011537020.1
XM_017019916.1 597 UTR 3 XP_016875405.1
XM_017019917.1 597 UTR 3 XP_016875406.1
Gene
HSP90B1
Gene Name
heat shock protein 90 beta family member 1
There are no transcripts associated with this gene.

View Full Product Details