Product Details

SNP ID
rs1109965
Assay Type
Functionally tested
NCBI dbSNP Submissions
46
Location
Chr.1:197913560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTCCACTGTCTGGAGAAAGGCC[C/T]CAGCGTGTCCCGGCAATCCGCTCAC
Phenotype
MIM: 606066
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf53 PubMed Links

Gene Details

Gene
C1orf53
Gene Name
chromosome 1 open reading frame 53
There are no transcripts associated with this gene.

Gene
LHX9
Gene Name
LIM homeobox 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014434.1 Intron NP_001014434.1
NM_020204.2 Intron NP_064589.2
XM_005245350.3 Intron XP_005245407.2
XM_011509781.2 Intron XP_011508083.2
XM_017001849.1 Intron XP_016857338.1

View Full Product Details