Product Details
- SNP ID
-
rs1045752
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:4251087 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAATTCAATGCTCGGAAGGAGAC[A/G]GTGCAGAACGAGGTCTACCTGGGCC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC94
PubMed Links
Gene Details
- Gene
- CCDC94
- Gene Name
- coiled-coil domain containing 94
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018074.4 |
219 |
Silent Mutation |
ACA,ACG |
T62T |
NP_060544.2 |
View Full Product Details