Product Details
- SNP ID
-
rs1042461
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:3632561 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GGCGGGAACATAGCTGCTGAGTGGC[C/T]GCAGTGCTTGGCAGGGCGAGGGCTG
- Phenotype
-
MIM: 606102
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CACTIN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3746124] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CACTIN
- Gene Name
- cactin, spliceosome C complex subunit
There are no transcripts associated with this gene.
- Gene
- PIP5K1C
- Gene Name
- phosphatidylinositol-4-phosphate 5-kinase type 1 gamma
View Full Product Details