Product Details

SNP ID
rs1628701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5892532 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTTGGGGCTGTATTGCCCATTC[C/T]TAGAAGCCATCACTTCCCGGCTGCG
Phenotype
MIM: 612638
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFA11 PubMed Links

Gene Details

Gene
NDUFA11
Gene Name
NADH:ubiquinone oxidoreductase subunit A11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193375.1 1377 UTR 3 NP_001180304.1
NM_175614.4 1377 Intron NP_783313.1

View Full Product Details