Product Details
- SNP ID
-
rs1799989
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:89177755 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCACTGGTGGGATACGAGCCAATT[C/A]GAAAGAAAAGTCAGTCATGTGCTTT
- Phenotype
-
MIM: 606933
- Polymorphism
- C/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TYR
PubMed Links
Gene Details
- Gene
- TYR
- Gene Name
- tyrosinase
View Full Product Details