Product Details

SNP ID
rs8322
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:42807145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAAACAATAGAAAAAGTGTTTT[G/T]TGTGTGTGTTTTTTTTTTTTTTTTA
Phenotype
MIM: 609017
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ERMAP PubMed Links

Gene Details

Gene
ERMAP
Gene Name
erythroblast membrane associated protein (Scianna blood group)
There are no transcripts associated with this gene.

Gene
LOC100129924
Gene Name
uncharacterized LOC100129924
There are no transcripts associated with this gene.

Gene
SVBP
Gene Name
small vasohibin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199342.3 705 UTR 3 NP_955374.1
XM_017001226.1 705 UTR 3 XP_016856715.1

View Full Product Details