Product Details
- SNP ID
-
rs993974
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:120238792 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGTCTAATGGAGGAGACAAATAGGT[A/G]AAGAGATTGATACAATCCAGAGTGA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NKAPP1
PubMed Links
Gene Details
- Gene
- NKAPP1
- Gene Name
- NFKB activating protein pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details