Product Details
- SNP ID
-
rs12305014
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:14974405 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGAAACTGCATAGGGATTAGTCGAC[C/T]GTTCCGGTTACCTAGTGCAAATCCT
- Phenotype
-
MIM: 601190
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PDE6H
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs61910465] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PDE6H
- Gene Name
- phosphodiesterase 6H
View Full Product Details