Product Details

SNP ID
rs8084936
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:46334475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGAGCGGTGGCTTCCCCCCACCCC[A/C]TTTTAGGGGGACCCCCGAGGGTGGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RNF165 PubMed Links

Gene Details

Gene
RNF165
Gene Name
ring finger protein 165
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256758.1 Intron NP_001243687.1
NM_152470.2 Intron NP_689683.2
XM_011526016.2 Intron XP_011524318.1
XM_017025788.1 Intron XP_016881277.1

View Full Product Details