Product Details

SNP ID
rs1500501
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:77540110 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATGTTCAGTATTATAGATTGATT[A/G]TCTCTGTTAACTTTATGTAATTTAT
Phenotype
MIM: 605149
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXCL13 PubMed Links

Gene Details

Gene
CXCL13
Gene Name
C-X-C motif chemokine ligand 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006419.2 Intron NP_006410.1
XM_006714063.3 Intron XP_006714126.1

View Full Product Details