Product Details
- SNP ID
-
rs400488
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:29975258 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GAGCCCCATAGTCCCCACAGCAATA[A/G]GAAGCACCAGCTCCTGGTCCCGAAA
- Phenotype
-
MIM: 615797
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
HCG9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139658941] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- HCG9
- Gene Name
- HLA complex group 9 (non-protein coding)
There are no transcripts associated with this gene.
View Full Product Details