Product Details

SNP ID
rs1150704
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:28216593 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTGGGTATTCCCTTTGCCCTTCAGA[G/T]CTAACCTATTTTTTTGGTGGGGGTG
Phenotype
MIM: 602246
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TOB2P1 PubMed Links

Gene Details

Gene
TOB2P1
Gene Name
transducer of ERBB2, 2 pseudogene 1
There are no transcripts associated with this gene.

Gene
ZSCAN9
Gene Name
zinc finger and SCAN domain containing 9
There are no transcripts associated with this gene.

View Full Product Details