Product Details

SNP ID
rs1795241
Assay Type
Validated
NCBI dbSNP Submissions
54
Location
Chr.1:171142226 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
GAGGTCATCTGGTGTGAGAGGGAGA[G/T]ACACATCTTGAATCCAGCAGCAACG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FMO6P PubMed Links

Gene Details

Gene
FMO6P
Gene Name
flavin containing monooxygenase 6 pseudogene
There are no transcripts associated with this gene.

View Full Product Details