Product Details

SNP ID
rs812383
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:232872422 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTCAGGGCCCTGTGGTCAGAACA[A/C]ACTTATTTACCAAAAGGCAGGGCGG
Phenotype
MIM: 605991
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C2orf82 PubMed Links

Gene Details

Gene
C2orf82
Gene Name
chromosome 2 open reading frame 82
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_206895.1 Intron NP_996778.1
XM_017004080.1 Intron XP_016859569.1
XM_017004081.1 Intron XP_016859570.1
XM_017004082.1 Intron XP_016859571.1
XM_017004083.1 Intron XP_016859572.1
XM_017004084.1 Intron XP_016859573.1
Gene
NGEF
Gene Name
neuronal guanine nucleotide exchange factor
There are no transcripts associated with this gene.

View Full Product Details