Product Details

SNP ID
rs1196865
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:105175708 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCAATGCTGTGGGGCATGTGATC[A/C]TACTCTGAGGAGCAAGGCCTTAGAG
Phenotype
MIM: 606231 MIM: 615748
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APPL2 PubMed Links

Gene Details

Gene
APPL2
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001251904.1 Intron NP_001238833.1
NM_001251905.1 Intron NP_001238834.1
NM_018171.3 Intron NP_060641.2
XM_006719472.1 Intron XP_006719535.1
XM_011538530.2 Intron XP_011536832.1
XM_011538531.2 Intron XP_011536833.1
XM_011538532.2 Intron XP_011536834.1
XM_017019551.1 Intron XP_016875040.1
XM_017019552.1 Intron XP_016875041.1
XM_017019553.1 Intron XP_016875042.1
XM_017019554.1 Intron XP_016875043.1
Gene
KIAA1033
Gene Name
KIAA1033
There are no transcripts associated with this gene.

View Full Product Details