Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001251904.1 | 4285 | UTR 3 | NP_001238833.1 | ||
NM_001251905.1 | 4285 | UTR 3 | NP_001238834.1 | ||
NM_018171.3 | 4285 | UTR 3 | NP_060641.2 | ||
XM_006719472.1 | 4285 | UTR 3 | XP_006719535.1 | ||
XM_011538530.2 | 4285 | UTR 3 | XP_011536832.1 | ||
XM_011538531.2 | 4285 | UTR 3 | XP_011536833.1 | ||
XM_011538532.2 | 4285 | UTR 3 | XP_011536834.1 | ||
XM_017019551.1 | 4285 | UTR 3 | XP_016875040.1 | ||
XM_017019552.1 | 4285 | UTR 3 | XP_016875041.1 | ||
XM_017019553.1 | 4285 | UTR 3 | XP_016875042.1 | ||
XM_017019554.1 | 4285 | UTR 3 | XP_016875043.1 |