Product Details

SNP ID
rs1804738
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:93121931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTGTCCTTGAGTGGGTCGAGACC[C/T]TTCTCTGGGCACAGGTCTCTGCAGC
Phenotype
MIM: 602062
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARD19 PubMed Links

Gene Details

Gene
CARD19
Gene Name
caspase recruitment domain family member 19
There are no transcripts associated with this gene.

Gene
NINJ1
Gene Name
ninjurin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004148.3 912 UTR 3 NP_004139.2
XM_011518716.1 912 UTR 3 XP_011517018.1

View Full Product Details