Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001290149.1 | 438 | UTR 5 | NP_001277078.1 | ||
NM_001290150.1 | 438 | UTR 5 | NP_001277079.1 | ||
NM_001290151.1 | 438 | UTR 5 | NP_001277080.1 | ||
NM_001290152.1 | 438 | UTR 5 | NP_001277081.1 | ||
NM_001290153.1 | 438 | UTR 5 | NP_001277082.1 | ||
NM_001290154.1 | 438 | UTR 5 | NP_001277083.1 | ||
NM_001290155.1 | 438 | UTR 5 | NP_001277084.1 | ||
NM_032712.2 | 438 | UTR 5 | NP_116101.1 | ||
NM_199249.2 | 438 | UTR 5 | NP_954857.1 | ||
NM_199250.2 | 438 | UTR 5 | NP_954858.1 |