Product Details

SNP ID
rs1268158
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:108754301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATTCTGAAGGTTCAGAAGCATGT[C/T]TTCTTTATCCATCCCCTGCCTCACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00222 PubMed Links

Gene Details

Gene
LINC00222
Gene Name
long intergenic non-protein coding RNA 222
There are no transcripts associated with this gene.

View Full Product Details