Product Details

SNP ID
rs1433108
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39539561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAATCGCCTGCGGCGTTCTTCAAG[C/T]TCTTGTATTCTGCCGGGAGCAATCG
Phenotype
MIM: 609773
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EID2 PubMed Links

Gene Details

Gene
EID2
Gene Name
EP300 interacting inhibitor of differentiation 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153232.3 638 Silent Mutation GAA,GAG E173E NP_694964.3
Gene
EID2B
Gene Name
EP300 interacting inhibitor of differentiation 2B
There are no transcripts associated with this gene.

View Full Product Details